LIF458786963 Life Chemicals Inc.

3-(chloromethyl)-1,2,4-oxadiazole

  • MDL number MFCD00052097
  • Cas Number : 51791-12-9
  • Molecular Weight : 118.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(chloromethyl)-1,2,4-oxadiazole
Inchi Code 1S/C3H3ClN2O/c4-1-3-5-2-7-6-3/h2H,1H2
Inchi Key YSNKGJCEHOJIDK-UHFFFAOYSA-N

Description

Purity 95%
Physical Form liquid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart