LIF458786969 Life Chemicals Inc.

(6-methoxybenzo[d][1,3]dioxol-5-yl)methanamine

  • MDL number MFCD16061312
  • Cas Number : 1250007-67-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart