LIF458786970 Life Chemicals Inc.

6-methoxybenzo[d][1,3]dioxole-5-carbonitrile

  • MDL number MFCD23709643
  • Cas Number : 1427365-61-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart