LIF458786971 Life Chemicals Inc.

O-(cyclopropylmethyl)hydroxylamine hydrochloride

  • MDL number MFCD08689925
  • Cas Number : 74124-04-2
  • Molecular Weight : 123.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name [(aminooxy)methyl]cyclopropane hydrochloride
Inchi Code 1S/C4H9NO.ClH/c5-6-3-4-1-2-4;/h4H,1-3,5H2;1H
Inchi Key HMMFZNBOQJOULD-UHFFFAOYSA-N

Description

Purity 95%
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart