LIF458786978 Life Chemicals Inc.

tert-butyl 3-(2-hydroxyethoxy)azetidine-1-carboxylate

  • MDL number MFCD28129779
  • Cas Number : 1146951-82-7
  • Molecular Weight : 217.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart