LIF458786980 Life Chemicals Inc.

2,4-dioxo-1,2,3,4-tetrahydroquinazoline-6-sulfonyl chloride

  • MDL number MFCD07186482
  • Cas Number : 56044-12-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart