LIF458786984 Life Chemicals Inc.

1-phenylpiperidin-3-ol

  • MDL number MFCD11109784
  • Cas Number : 80710-25-4
  • Molecular Weight : 177.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart