LIF458786985 Life Chemicals Inc.

5-methyl-1,3-diphenyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD00510082
  • Cas Number : 15409-48-0
  • Molecular Weight : 278.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-methyl-1,3-diphenyl-1H-pyrazole-4-carboxylic acid
Inchi Code 1S/C17H14N2O2/c1-12-15(17(20)21)16(13-8-4-2-5-9-13)18-19(12)14-10-6-3-7-11-14/h2-11H,1H3,(H,20,21)
Inchi Key FWIYNXGMTSTQBJ-UHFFFAOYSA-N

Description

Purity 95%
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart