LIF458786986 Life Chemicals Inc.

(4-phenylmorpholin-2-yl)methanamine

  • MDL number MFCD06738826
  • Cas Number : 112913-99-2
  • Molecular Weight : 192.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-phenyl-2-morpholinyl)methanamine
Inchi Code 1S/C11H16N2O/c12-8-11-9-13(6-7-14-11)10-4-2-1-3-5-10/h1-5,11H,6-9,12H2
Inchi Key YAKKATWFMLCGMI-UHFFFAOYSA-N

Description

Purity 95%
Physical Form oil
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart