LIF458786996 Life Chemicals Inc.

2-(aminomethyl)-1,2,3,4-tetrahydronaphthalen-2-ol

  • MDL number MFCD12152094
  • Cas Number : 1179255-66-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form oil
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart