LIF458787001 Life Chemicals Inc.

7-fluorobenzo[b]thiophen-3(2H)-one

  • MDL number MFCD28630476
  • Cas Number : 1558342-18-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart