LIF458787003 Life Chemicals Inc.

2-(3-methyl-1H-pyrazol-5-yl)ethan-1-amine

  • MDL number MFCD01543581
  • Cas Number : 54055-40-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form oil
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart