LIF458787007 Life Chemicals Inc.

5,7-dibromo-1H-indazole

  • MDL number MFCD05664000
  • Cas Number : 50477-28-6
  • Molecular Weight : 275.93

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart