LIF458787011 Life Chemicals Inc.

(1-methoxycyclohexyl)methanamine

  • MDL number MFCD16681072
  • Cas Number : 90886-41-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart