LIF458787012 Life Chemicals Inc.

ethyl N-(1-amino-4-methylpentan-2-yl)carbamate

  • MDL number MFCD11099482
  • Cas Number : 1087792-22-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart