LIF458787015 Life Chemicals Inc.

3-(pyridin-3-yloxy)aniline

  • MDL number MFCD14583123
  • Cas Number : 116289-71-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart