LIF458787017 Life Chemicals Inc.

2-bromo-5-(oxan-4-yl)-1,3,4-thiadiazole

  • MDL number MFCD18345168
  • Cas Number : 1340436-02-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart