LIF458787018 Life Chemicals Inc.

4-bromopentanenitrile

  • MDL number MFCD21184651
  • Cas Number : 14470-12-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form oil
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart