LIF458787019 Life Chemicals Inc.

2-(methoxymethoxy)aniline

  • MDL number MFCD21195210
  • Cas Number : 88733-56-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart