LIF458787020 Life Chemicals Inc.

3-methoxy-4-(pyridin-3-yloxy)aniline

  • MDL number MFCD24639675
  • Cas Number : 871020-12-1
  • Molecular Weight : 216.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart