LIF458787026 Life Chemicals Inc.

2-(1-methyl-1H-pyrazol-5-yl)cyclopropane-1-carboxylic acid

  • MDL number MFCD28133323
  • Cas Number : 1601247-09-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart