LIF458787032 Life Chemicals Inc.

3-methoxybutan-1-amine hydrochloride

  • MDL number MFCD22422012
  • Cas Number : 1417569-76-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form oil
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart