LIF458787036 Life Chemicals Inc.

2-chloro-4-(pyridin-3-yloxy)aniline

  • MDL number MFCD26400254
  • Cas Number : 315227-20-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart