LIF458787038 Life Chemicals Inc.

2'-(trifluoromethyl)-[1,1'-biphenyl]-4-carbaldehyde

  • MDL number MFCD01631916
  • Cas Number : 198205-95-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart