LIF459291131 Life Chemicals Inc.

(S)-1,3-dimethyl-2-oxoimidazolidine-4-carboxylic acid

  • MDL number MFCD29046409
  • Cas Number : 1069090-20-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart