LIF459291132 Life Chemicals Inc.

2-(pyrrolidin-3-yl)thiazole dihydrochloride

  • MDL number MFCD29046561
  • Cas Number : 1820618-32-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart