LIF459291137 Life Chemicals Inc.

3-(chloromethyl)-6,7-dihydro-5H-pyrrolo[2,1-c][1,2,4]triazole hydrochloride

  • MDL number MFCD29046932
  • Cas Number : 1820685-05-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart