LIF459291138 Life Chemicals Inc.

3-((benzyl(ethyl)amino)methyl)aniline

  • MDL number MFCD10021240
  • Cas Number : 1018517-42-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart