LIF459291141 Life Chemicals Inc.

4-chloro-5,6-dimethylpyrimidine

  • MDL number MFCD09909775
  • Cas Number : 67434-65-5
  • Molecular Weight : 142.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-chloro-5,6-dimethylpyrimidine
Inchi Code 1S/C6H7ClN2/c1-4-5(2)8-3-9-6(4)7/h3H,1-2H3
Inchi Key MJRSZADMKJVGQV-UHFFFAOYSA-N

Description

Purity 95%
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart