LIF459291143 Life Chemicals Inc.

2-(tert-butyl)-5-methyl-2,4-dihydro-3H-pyrazol-3-one

  • MDL number MFCD00828688
  • Cas Number : 132214-71-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart