LIF459291148 Life Chemicals Inc.

2-chloro-6,7-dimethoxyquinoxaline

  • MDL number MFCD09836153
  • Cas Number : 216699-86-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart