LIF459291150 Life Chemicals Inc.

thiazolo[4,5-b]pyrazin-2-amine

  • MDL number MFCD15526540
  • Cas Number : 112342-71-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart