LIF459291152 Life Chemicals Inc.

2-((2-oxo-2H-chromen-4-yl)oxy)acetic acid

  • MDL number MFCD09837287
  • Cas Number : 462094-45-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart