LIF459291159 Life Chemicals Inc.

2-cyclohexylpyrimidine-5-carbaldehyde

  • MDL number MFCD09864503
  • Cas Number : 959240-13-2
  • Molecular Weight : 190.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-cyclohexyl-5-pyrimidinecarbaldehyde
Inchi Code 1S/C11H14N2O/c14-8-9-6-12-11(13-7-9)10-4-2-1-3-5-10/h6-8,10H,1-5H2
Inchi Key FDLLJEWSIWXXEG-UHFFFAOYSA-N

Description

Purity 95%
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart