LIF459291163 Life Chemicals Inc.

tert-butyl 4-amino-2-fluorobenzoate

  • MDL number MFCD06658385
  • Cas Number : 140373-77-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart