LIF459291166 Life Chemicals Inc.

1-methyl-2-oxo-1,2-dihydroquinoline-3-carbaldehyde

  • MDL number MFCD06738236
  • Cas Number : 67735-60-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart