LIF459291175 Life Chemicals Inc.

tert-butyl (2-aminopropyl)carbamate

  • MDL number MFCD09788609
  • Cas Number : 255735-88-7
  • Molecular Weight : 174.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart