LIF459291177 Life Chemicals Inc.

1-(3,5-dimethyl-1H-pyrazol-1-yl)propan-2-one

  • MDL number MFCD03305776
  • Cas Number : 361343-66-0
  • Molecular Weight : 152.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(3,5-dimethyl-1H-pyrazol-1-yl)acetone
Inchi Code 1S/C8H12N2O/c1-6-4-7(2)10(9-6)5-8(3)11/h4H,5H2,1-3H3
Inchi Key ROLPGYNPJZZIGQ-UHFFFAOYSA-N

Description

Purity 95%
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart