LIF459291180 Life Chemicals Inc.

methyl N'-cyanocarbamimidothioate

  • MDL number MFCD02181250
  • Cas Number : 860354-94-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart