LIF459291181 Life Chemicals Inc.

3-bromo-5-(oxan-4-yl)-1,2,4-thiadiazole

  • MDL number MFCD28151670
  • Cas Number : 1600307-14-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart