LIF459291182 Life Chemicals Inc.

4-chloro-2-methyl-6-(trifluoromethyl)pyrimidine

  • MDL number MFCD03265399
  • Cas Number : 5993-98-6
  • Molecular Weight : 196.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart