LIF459291187 Life Chemicals Inc.

1,3,4-Pyrrolidinetricarboxylic acid, 1-(1,1-dimethylethyl) 3,4-diethyl ester, (3R,4S)-rel-

  • MDL number MFCD21602371
  • Cas Number : 1360547-54-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart