LIF465533719 Life Chemicals Inc.

(S)-2-oxoimidazolidine-4-carboxylic acid

  • MDL number MFCD00272206
  • Cas Number : 41371-53-3
  • Molecular Weight : 130.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart