LIF465533734 Life Chemicals Inc.

1-(2-bromoethyl)-1H-pyrazole

  • MDL number MFCD03727235
  • Cas Number : 119291-22-4
  • Molecular Weight : 175.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart