LIF466082892 Life Chemicals Inc.

5-(aminomethyl)-1,2-dihydro-4H-pyrrolo[3,2,1-ij]quinolin-4-one hydrochloride

  • MDL number MFCD29090789
  • Cas Number : 1881328-51-3
  • Molecular Weight : 236.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart