LIF466082893 Life Chemicals Inc.

6-(aminomethyl)-2,3-dihydro-1H,5H-pyrido[3,2,1-ij]quinolin-5-one hydrochloride

  • MDL number MFCD29090790
  • Cas Number : 1881322-03-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart