LIF466082894 Life Chemicals Inc.

(1-ethoxycyclobutyl)methanamine hydrochloride

  • MDL number MFCD29090761
  • Cas Number : 1881331-31-2
  • Molecular Weight : 165.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart