LIF466082895 Life Chemicals Inc.

2-(5-methyl-1H-pyrazol-3-yl)morpholine dihydrochloride

  • MDL number MFCD30011207
  • Cas Number : 2034153-26-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart