LIF466082912 Life Chemicals Inc.

4-(chloromethyl)-2-cyclopropylthiazole hydrochloride

  • MDL number MFCD29090791
  • Cas Number : 135207-10-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart