LIF466082913 Life Chemicals Inc.

ethyl 2-oxo-2-(4-oxooxan-3-yl)acetate

  • MDL number MFCD21604042
  • Cas Number : 857177-01-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart